Thiophen-3-acetonitril
Lieferant: Molekula
Synonyme:
2-(Thiophen-3-yl)acetonitrile, 3-Thiopheneacetonitrile
46123285-25G
MOLE46123285-25GEA
0
EUR
MOLE46123285-25G
Thiophen-3-acetonitril
Thiophen-3-acetonitril
Formel:
C₆H₅NS Molecular Weight: 123,18 g/mol |
MDL:
MFCD00005471 CAS-Nummer: 13781-53-8 EINECS: 237-433-1 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...