Ethyl-3-piperidincarboxylat
Lieferant: Molekula
Synonyme:
Ethyl nipecotate, Ethyl 3-Piperidinecarboxylate, Ethyl piperidine-3-carboxylate
46246962-100G
MOLE46246962-100GEA
71.6
EUR
MOLE46246962-100G
Ethyl-3-piperidincarboxylat
Ethyl-3-piperidincarboxylat
Formel:
C₈H₁₅NO₂ Molecular Weight: 157,21 g/mol |
MDL:
MFCD00005991 CAS-Nummer: 5006-62-2 EINECS: 225-681-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...