N,N-Dimethylformamiddimethylacetal
Lieferant: Molekula
|
Gefahr
|
Synonyme:
1,1-Dimethoxytrimethylamin
46666234-100G
MOLE46666234-100GEA
89.7
EUR
MOLE46666234-100G
N,N-Dimethylformamiddimethylacetal
N,N-Dimethylformamiddimethylacetal
Formel:
C₅H₁₃NO₂ Molecular Weight: 119,16 g/mol Siedepunkt: 102 °C (1013 hPa) Dichte: 0,893 g/cm³ (20 °C) Flammpunkt: 6 °C |
MDL:
MFCD00008482 CAS-Nummer: 4637-24-5 EINECS: 225-063-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...