4-Nitroimidazol

Lieferant: Molekula
Achtung

Synonyme: 4-Nitro-1H-imidazol

46670097-25G
MOLE46670097-25GEA 0 EUR
MOLE46670097-25G
4-Nitroimidazol
4-Nitroimidazol
Formel: C₃H₃N₃O₂
Molecular Weight: 113,08 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005196
CAS-Nummer: 3034-38-6
EINECS: 221-224-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR