2,4-Diaminoanisolsulfat Hydrat

Lieferant: Molekula

46715302-500G
MOLE46715302-500GEA 0 EUR
MOLE46715302-500G
2,4-Diaminoanisolsulfat Hydrat
2,4-Diaminoanisolsulfat Hydrat
Formel: C₇H₁₂N₂O₅S·H₂O
MDL: MFCD00012978
CAS-Nummer: 123333-56-2
EINECS: 254-323-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR