Dibenzo-24-krone-8 (Dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosan)

Lieferant: Molekula

Synonyme: Dibenzo-24-crown-8

46817380-1G
MOLE46817380-1GEA 0 EUR
MOLE46817380-1G
Dibenzo-24-krone-8 (Dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosan)
Dibenzo-24-krone-8 (Dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosan)
Formel: C₂₄H₃₂O₈
Molecular Weight: 448,51 g/mol
MDL: MFCD00005101
CAS-Nummer: 14174-09-5
EINECS: 238-027-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR