(R)-(+)-1,1'-Bi-2-naphthol

Lieferant: Molekula

Synonyme: (R)-[1,1'-Binaphthalene]-2,2'-diol, (R)-BINOL, (R)-(+)-1,1'-Binaphthyl-2,2'-Diol

47008781-1G
MOLE47008781-1GEA 0 EUR
MOLE47008781-1G
(R)-(+)-1,1'-Bi-2-naphthol
(R)-(+)-1,1'-Bi-2-naphthol
Formel: C₂₀H₁₄O₂
Molecular Weight: 286,33 g/mol
MDL: MFCD00004068
CAS-Nummer: 18531-94-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR