3,5-Dimethyl-4-hydroxybenzonitril

Lieferant: Molekula

Synonyme: 4-Hydroxy-3,5-dimethylbenzonitrile

47173898-1G
MOLE47173898-1GEA 0 EUR
MOLE47173898-1G
3,5-Dimethyl-4-hydroxybenzonitril
3,5-Dimethyl-4-hydroxybenzonitril
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
MDL: MFCD00221716
CAS-Nummer: 4198-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR