D(-)-Penicillamin

Lieferant: Molekula

47221894-5G
MOLE47221894-5GEA 0 EUR
MOLE47221894-5G
D(-)-Penicillamin
D(-)-Penicillamin
Formel: C₅H₁₁NO₂S
MDL: MFCD00064302
CAS-Nummer: 52-67-5
EINECS: 200-148-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR