HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)
Lieferant: Molekula
Synonyme:
HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
47376574-100G
MOLE47376574-100GEA
116
EUR
MOLE47376574-100G
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)
Formel:
C₈H₁₈N₂O₄S Molecular Weight: 238.31 g/mol Schmelzpunkt: 210…215 °C Dichte: 1.448 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006158 CAS-Nummer: 7365-45-9 EINECS: 230-907-9 Merck Index: 13,04672 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...