4-Nitrophenethylbromid
Lieferant: Molekula
Synonyme:
2-(4-Nitrophenyl)-1-bromoethane, 1-(2-Bromoethyl)-4-nitrobenzene
47797515-25G
MOLE47797515-25GEA
0
EUR
MOLE47797515-25G
4-Nitrophenethylbromid
4-Nitrophenethylbromid
Formel:
C₈H₈BrNO₂ Molecular Weight: 230,06 g/mol |
MDL:
MFCD00007386 CAS-Nummer: 5339-26-4 EINECS: 226-271-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...