2,3,5,6-Tetramethyl-m-phenylendiamin
Lieferant: Molekula
Synonyme:
2,3,5,6-Tetramethyl-1,3-phenylendiamin, 1,4-Diamino-2,3,5,6-tetramethylbenzol, 2,3,5,6-Tetramethylbenzol-1,3-diamin
47973115-5G
MOLE47973115-5GEA
0
EUR
MOLE47973115-5G
2,3,5,6-Tetramethyl-m-phenylendiamin
2,3,5,6-Tetramethyl-m-phenylendiamin
Formel:
C₁₀H₁₆N₂ Molecular Weight: 164,25 g/mol Siedepunkt: 316 °C (1013 hPa) Schmelzpunkt: 150…153 °C |
MDL:
MFCD00007905 CAS-Nummer: 3102-87-2 EINECS: 221-457-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...