3-Brombenzylalkohol
Lieferant: Molekula
Synonyme:
(3-Bromophenyl)methanol
47984076-5G
MOLE47984076-5GEA
86
EUR
MOLE47984076-5G
3-Brombenzylalkohol
3-Brombenzylalkohol
Formel:
C₇H₇BrO Molecular Weight: 187,04 g/mol Siedepunkt: 165 °C (1013 hPa) Dichte: 1,56 g/cm³ (20 °C) |
MDL:
MFCD00004629 CAS-Nummer: 15852-73-0 EINECS: 239-975-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...