(1H-Imidazol-4-yl)methanol Hydrochlorid

Lieferant: Molekula

Synonyme: 4-Imidazolemethanol hydrochloride, 4(5)-Imidazolemethanol hydrochloride, 4-(Hydroxymethyl)imidazole hydrochloride

48181383-5G
MOLE48181383-5GEA 0 EUR
MOLE48181383-5G
(1H-Imidazol-4-yl)methanol Hydrochlorid
(1H-Imidazol-4-yl)methanol Hydrochlorid
Formel: C₄H₇ClN₂O
Molecular Weight: 134,57 g/mol
MDL: MFCD00012697
CAS-Nummer: 32673-41-9
EINECS: 251-150-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR