Gemfibrozil

Lieferant: Molekula

Synonyme: CI-719, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeriansäure, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentansäure

48653561-5G
MOLE48653561-5GEA 101 EUR
MOLE48653561-5G
Gemfibrozil
Gemfibrozil
Formel: C₁₅H₂₂O₃
Molecular Weight: 250,34 g/mol
Siedepunkt: 395 °C (1013 hPa)
Schmelzpunkt: 59…61 °C
MDL: MFCD00079335
CAS-Nummer: 25812-30-0
EINECS: 247-280-2
Merck Index: 12,04394

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR