5-Chlor-2-fluoranilin

Lieferant: Molekula
Achtung

48709208-25G
MOLE48709208-25GEA 0 EUR
MOLE48709208-25G
5-Chlor-2-fluoranilin
5-Chlor-2-fluoranilin
Formel: C₆H₅ClFN
MDL: MFCD00069416
CAS-Nummer: 2106-05-0
EINECS: 218-284-1
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR