4-Mercaptobenzoesäure

Lieferant: Molekula

Synonyme: 4-Thiobenzoic acid

49383024-5G
MOLE49383024-5GEA 0 EUR
MOLE49383024-5G
4-Mercaptobenzoesäure
4-Mercaptobenzoic acid
Formel: C₇H₆O₂S
MDL: MFCD00016617
CAS-Nummer: 1074-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR