2-Amino-6-nitrobenzothiazol
Lieferant: Molekula
Synonyme:
6-Nitro-1,3-benzothiazol-2-amin, 6-Nitrobenzothiazol-2-ylamin
49445000-25G
MOLE49445000-25GEA
0
EUR
MOLE49445000-25G
2-Amino-6-nitrobenzothiazol
2-Amino-6-nitrobenzothiazole
Formel:
C₇H₅N₃O₂S Molecular Weight: 195,2 g/mol Schmelzpunkt: 252 °C Dichte: 1,649 g/cm³ (20 °C) |
MDL:
MFCD00005786 CAS-Nummer: 6285-57-0 EINECS: 228-513-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...