(R)-(+)-2-Hydroxy-3-phenylpropionsäure
Lieferant: Molekula
Synonyme:
D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-(+)-Phenyllactic acid
49727033-1G
MOLE49727033-1GEA
0
EUR
MOLE49727033-1G
(R)-(+)-2-Hydroxy-3-phenylpropionsäure
(R)-(+)-2-Hydroxy-3-phenylpropionsäure
Formel:
C₉H₁₀O₃ |
MDL:
MFCD00078062 CAS-Nummer: 7326-19-4 EINECS: 230-803-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...