ω-Brom-2-fluoracetophenon
Lieferant: Molekula
Synonyme:
2-Brom-2'-fluoracetophenon, 2-Brom-1-(2-fluorphenyl)ethan-1-on, α-Brom-2-fluoracetophenon, 2-Fluorphenacylbromid, Brommethyl-o-fluorphenylketon
50038779-1G
MOLE50038779-1GEA
0
EUR
MOLE50038779-1G
ω-Brom-2-fluoracetophenon
ω-Brom-2-fluoracetophenon
Formel:
C₈H₆BrFO Molecular Weight: 217,04 g/mol |
MDL:
MFCD00278796 CAS-Nummer: 655-15-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...