5-(Ethylthio)-1H-tetrazol (ETT)

Lieferant: Molekula

51057635-25G
MOLE51057635-25GEA 0 EUR
MOLE51057635-25G
5-(Ethylthio)-1H-tetrazol (ETT)
5-(Ethylthio)-1H-tetrazol (ETT)
Formel: C₃H₆N₄S
MDL: MFCD00068733
CAS-Nummer: 89797-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR