1,3-Dichlor-5-nitrobenzol
Lieferant: Molekula
|
Achtung
|
Synonyme:
3,5-Dichloronitrobenzene, 1,3-Dichloro-5-nitrobenzene
51470062-25G
MOLE51470062-25GEA
0
EUR
MOLE51470062-25G
1,3-Dichlor-5-nitrobenzol
1,3-Dichlor-5-nitrobenzol
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192 g/mol Schmelzpunkt: 64…65 °C Dichte: 1.69 g/cm³ (14 °C) |
MDL:
MFCD00007211 CAS-Nummer: 618-62-2 EINECS: 210-557-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...