Eriochromschwarz T
Lieferant: Molekula
|
Achtung
|
Synonyme:
1-(2-Hydroxy-1-naphthylazo)-6-nitro-2-naphthol-4-sulfonsäure Natriumsalz, Natrium-3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalinsulfonat
51499212-25G
MOLE51499212-25GEA
96.5
EUR
MOLE51499212-25G
Eriochromschwarz T
Eriochromschwarz T
Formel:
C₂₀H₁₂N₃NaO₇S Molecular Weight: 461,39 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003935 CAS-Nummer: 1787-61-7 EINECS: 217-250-3 UN: 3077 ADR: 9,III Merck Index: 13,03702 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...