5-(4-Pyridyl)-1H-tetrazol

Lieferant: Molekula

Synonyme: 4-(1H-Tetrazol-5-yl)pyridine

51928449-1G
MOLE51928449-1GEA 0 EUR
MOLE51928449-1G
5-(4-Pyridyl)-1H-tetrazol
5-(4-Pyridyl)-1H-tetrazole
Formel: C₆H₅N₅
Molecular Weight: 147,14 g/mol
MDL: MFCD00068115
CAS-Nummer: 14389-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR