2-Mercapto-1,3,4-thiadiazol
Lieferant: Molekula
Synonyme:
1,3,4-Thiadiazole-2-thiol
51938931-25G
MOLE51938931-25GEA
0
EUR
MOLE51938931-25G
2-Mercapto-1,3,4-thiadiazol
2-Mercapto-1,3,4-thiadiazole
Formel:
C₂H₂N₂S₂ Molecular Weight: 118,17 g/mol |
MDL:
MFCD00040404 CAS-Nummer: 18686-82-3 EINECS: 242-504-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...