5-Chlor-2-methoxybenzoesäure
Lieferant: Molekula
Synonyme:
5-Chlor-o-anissäure
52004574-500G
MOLE52004574-500GEA
0
EUR
MOLE52004574-500G
5-Chlor-2-methoxybenzoesäure
5-Chlor-2-methoxybenzoesäure
Formel:
C₈H₇ClO₃ Molecular Weight: 186,59 g/mol |
MDL:
MFCD00017546 CAS-Nummer: 3438-16-2 EINECS: 222-343-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...