Tetrapropylammoniumbromid
Lieferant: Molekula
Synonyme:
TPABr
52126738-100G
MOLE52126738-100GEA
0
EUR
MOLE52126738-100G
Tetrapropylammoniumbromid
Tetrapropylammoniumbromid
Formel:
C₁₂H₂₈BrN Molecular Weight: 266,27 g/mol Schmelzpunkt: 265…274 °C |
MDL:
MFCD00011840 CAS-Nummer: 1941-30-6 EINECS: 217-727-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...