2-Amino-6-(trifluormethyl)pyridin
Lieferant: Molekula
52130800-1G
MOLE52130800-1GEA
0
EUR
MOLE52130800-1G
2-Amino-6-(trifluormethyl)pyridin
2-Amino-6-(trifluoromethyl)pyridine
Formel:
C₆H₅F₃N₂ Molecular Weight: 162,11 g/mol Siedepunkt: 200 °C (1013 hPa) Schmelzpunkt: 78…80 °C |
MDL:
MFCD03093868 CAS-Nummer: 34486-24-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...