2'-Desoxycytidin

Lieferant: Molekula

Synonyme: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, deoxyC

52321233-1G
MOLE52321233-1GEA 71.6 EUR
MOLE52321233-1G
2'-Desoxycytidin
2'-Desoxycytidin
Formel: C₉H₁₃N₃O₄
Molecular Weight: 227,22 g/mol
MDL: MFCD00006547
CAS-Nummer: 951-77-9
EINECS: 213-454-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR