Ethyl-3-amino-1H-pyrazol-4-carboxylat
Lieferant: Molekula
Synonyme:
3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate
52361395-5G
MOLE52361395-5GEA
0
EUR
MOLE52361395-5G
Ethyl-3-amino-1H-pyrazol-4-carboxylat
Ethyl-3-amino-1H-pyrazol-4-carboxylat
Formel:
C₆H₉N₃O₂ Molecular Weight: 155,16 g/mol |
MDL:
MFCD00005238 CAS-Nummer: 6994-25-8 EINECS: 230-262-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...