Phloroglucin Dihydrat
Lieferant: Molekula
|
Achtung
|
Synonyme:
1,3,5-Trihydroxybenzol Dihydrat, 1,3,5-Benzoltriol Dihydrat
53027361-100G
MOLE53027361-100GEA
109
EUR
MOLE53027361-100G
Phloroglucin Dihydrat
Phloroglucin Dihydrat
Formel:
C₆H₆O₃·2H₂O Molecular Weight: 162,14 g/mol Schmelzpunkt: 218…221 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149090 CAS-Nummer: 6099-90-7 EINECS: 203-611-2 Merck Index: 13,07413 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...