1-Acetyl-4-piperidon

Lieferant: Molekula

Synonyme: N-Acetyl-4-piperidone

53288356-25G
MOLE53288356-25GEA 0 EUR
MOLE53288356-25G
1-Acetyl-4-piperidon
1-Acetyl-4-piperidon
Formel: C₇H₁₁NO₂
Molecular Weight: 141.17 g/mol
MDL: MFCD00006190
CAS-Nummer: 32161-06-1
EINECS: 250-937-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR