5-Aminoindol
Lieferant: Molekula
Synonyme:
5-Indolamin, Indol-5-ylamin, 5-Amino-1H-indol
53302534-1G
MOLE53302534-1GEA
0
EUR
MOLE53302534-1G
5-Aminoindol
5-Aminoindol
Formel:
C₈H₈N₂ Molecular Weight: 132,16 g/mol Siedepunkt: 190 °C (6 mmHg) Schmelzpunkt: 131…134 °C |
MDL:
MFCD00005679 CAS-Nummer: 5192-03-0 EINECS: 225-977-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...