Eriochromcyanin R

Lieferant: Molekula

Synonyme: Chromoxane Cyanine R

53361433
MOLE53361433EA 0 EUR
MOLE53361433
Eriochromcyanin R
Eriochromcyanin R
Formel: C₂₃H₁₅Na₃O₉S
MDL: MFCD00012411
CAS-Nummer: 3564-18-9
EINECS: 222-641-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR