Cetrimoniumchlorid
Lieferant: Molekula
Synonyme:
Hexadecyltrimethylammoniumchlorid, Cetyltrimethylammoniumchlorid, CTAC, N-Cetyl-N,N,N-trimethylammoniumchlorid, N-Hexadecyl-N,N,N-trimethylammoniumchlorid
53473813-100G
MOLE53473813-100GEA
160
EUR
MOLE53473813-100G
Cetrimoniumchlorid
Cetrimoniumchlorid
Formel:
C₁₉H₄₂ClN Molecular Weight: 320 g/mol Schmelzpunkt: 232…233 °C Dichte: 0,9 g/cm³ (20 °C) |
MDL:
MFCD00011773 CAS-Nummer: 112-02-7 EINECS: 203-928-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...