4-Methoxybenzolsulfonylchlorid
Lieferant: Molekula
Synonyme:
4-Methoxybenzolsulfonsäurechlorid, 4-Methoxybenzol-1-sulfonylchlorid, 4-Methoxybenzolsulfochlorid
53721304-100G
MOLE53721304-100GEA
0
EUR
MOLE53721304-100G
4-Methoxybenzolsulfonylchlorid
4-Methoxybenzolsulfonylchlorid
Formel:
C₇H₇ClO₃S Molecular Weight: 206,65 g/mol Siedepunkt: 316 °C (1013 hPa) Schmelzpunkt: 40…42 °C |
MDL:
MFCD00007446 CAS-Nummer: 98-68-0 EINECS: 202-692-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...