Kaliumhydrogenorthoarsenat Dihydrat

Lieferant: Molekula
Gefahr

54082809-5G
MOLE54082809-5GEA 0 EUR
MOLE54082809-5G
Kaliumhydrogenorthoarsenat Dihydrat
Kaliumhydrogenorthoarsenat Dihydrat
Formel: HAsK₂O₄·2H₂O
CAS-Nummer: 308103-48-2
UN: 1677
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR