(S)-(+)-1-Methoxy-2-propanol

Lieferant: Molekula

Synonyme: (S)-(+)-Propylenglycolmonomethylether

54373270-1G
MOLE54373270-1GEA 0 EUR
MOLE54373270-1G
(S)-(+)-1-Methoxy-2-propanol
(S)-(+)-1-Methoxy-2-propanol
Formel: C₄H₁₀O₂
Molecular Weight: 90,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632588
CAS-Nummer: 26550-55-0
UN: 3092
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR