L(+)-Lysin Monohydrat
Lieferant: Molekula
Synonyme:
K Monohydrat, (S)-(+)-2,6-Diaminohexansäure Monohydrat, H-Lys-OH.H2O, (S)-(+)-Lysin Monohydrat, (S)-(+)-2,6-Diaminocapronsäure Monohydrat, Lys Monohydrat
54686501-100G
MOLE54686501-100GEA
242
EUR
MOLE54686501-100G
L(+)-Lysin Monohydrat
L(+)-Lysin Monohydrat
Formel:
H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O Molecular Weight: 164,2 g/mol Schmelzpunkt: 225 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00151035 CAS-Nummer: 39665-12-8 EINECS: 200-294-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...