2-Thiophencarbonitril
Lieferant: Molekula
Synonyme:
2-Thienylcyanid, Thiophen-2-carbonsäurenitril, Thiophen-2-carbonitril, 2-Cyanthiophen, 2-Thiophencarbonsäurenitril
54939601-25G
MOLE54939601-25GEA
0
EUR
MOLE54939601-25G
2-Thiophencarbonitril
2-Thiophencarbonitril
Formel:
C₅H₃NS Molecular Weight: 109,15 g/mol Siedepunkt: 190…192 °C (1013 hPa) Dichte: 1,18 g/cm³ (15 °C) |
MDL:
MFCD00005416 CAS-Nummer: 1003-31-2 EINECS: 213-706-0 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...