Dodecyltrimethylammoniumchlorid 25% in wässriger Lösung
Lieferant: Molekula
|
Achtung
|
Synonyme:
(1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC
54952018-1L
MOLE54952018-1LEA
0
EUR
MOLE54952018-1L
Dodecyltrimethylammoniumchlorid 25% in wässriger Lösung
Dodecyltrimethylammonium chloride
Formel:
C₁₅H₃₄ClN Molecular Weight: 263,89 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041974 CAS-Nummer: 112-00-5 EINECS: 203-927-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...