2-Chinoxalincarbonsäure

Lieferant: Molekula

55827735-500MG
MOLE55827735-500MGEA 0 EUR
MOLE55827735-500MG
2-Chinoxalincarbonsäure
2-Quinoxalinecarboxylic acid
Formel: C₉H₆N₂O₂
Molecular Weight: 174,16 g/mol
MDL: MFCD00012334
CAS-Nummer: 879-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR