Isonicotinaldehydoxim

Lieferant: Molekula

Synonyme: 4-Pyridinecarbaldehyde oxime, Pyridine-4-carboxaldoxime

55953844-100G
MOLE55953844-100GEA 291 EUR
MOLE55953844-100G
Isonicotinaldehydoxim
Isonicotinaldehydoxim
Formel: C₆H₆N₂O
Molecular Weight: 122,13 g/mol
MDL: MFCD00006446
CAS-Nummer: 696-54-8
EINECS: 211-796-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR