Butamben (Butyl-4-aminobenzoat)

Lieferant: Molekula

Synonyme: Butamben, Butyl 4-aminobenzoate

55959005-5G
MOLE55959005-5GEA 0 EUR
MOLE55959005-5G
Butamben (Butyl-4-aminobenzoat)
Butamben (Butyl-4-aminobenzoat)
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,25 g/mol
MDL: MFCD00017112
CAS-Nummer: 94-25-7
EINECS: 202-317-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR