6-Brom-2-naphthoesäure

Lieferant: Molekula

56202798-25G
MOLE56202798-25GEA 0 EUR
MOLE56202798-25G
6-Brom-2-naphthoesäure
6-Brom-2-naphthoesäure
Formel: C₁₁H₇BrO₂
Molecular Weight: 251,08 g/mol
MDL: MFCD01075720
CAS-Nummer: 5773-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR