4,4'-Dichlorbenzhydrol

Lieferant: Molekula

Synonyme: 4,4'-Dichlorobenzhydryl alcohol, DBH, 4,4'-Dichlorophenyl phenyl carbinol, 4,4'-Dichloro-α-phenylbenzyl alcohol, 4,4'-Dichlorodiphenylmethanol, Bis(4-chlorophenyl)methanol, Bis(4-chlorophenyl)carbinol

56493615-100G
MOLE56493615-100GEA 262 EUR
MOLE56493615-100G
4,4'-Dichlorbenzhydrol
4,4'-Dichlorbenzhydrol
Formel: C₁₃H₁₀Cl₂O
Molecular Weight: 253.13 g/mol
MDL: MFCD00000629
CAS-Nummer: 90-97-1
EINECS: 202-029-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR