(R)-(+)-Indolin-2-carbonsäure

Lieferant: Molekula

56971128-5G
MOLE56971128-5GEA 0 EUR
MOLE56971128-5G
(R)-(+)-Indolin-2-carbonsäure
(R)-(+)-Indolin-2-carbonsäure
Formel: C₉H₉NO₂
MDL: MFCD00792496
CAS-Nummer: 98167-06-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR