3-Methoxyzimtsäure

Lieferant: Molekula
Achtung

Synonyme: 3-(3-Methoxyphenyl)acrylic acid

57097526-25G
MOLE57097526-25GEA 0 EUR
MOLE57097526-25G
3-Methoxyzimtsäure
3-Methoxyzimtsäure
Formel: C₁₀H₁₀O₃
Molecular Weight: 178.19 g/mol
MDL: MFCD00004386
CAS-Nummer: 6099-04-3
EINECS: 228-049-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR