(p-Toluolsulfonyl)methylisocyanid

Lieferant: Molekula

Synonyme: 1-((Isocyanomethyl)sulphonyl)-4-methylbenzene, Tosylmethyl isocyanide

57251431-25G
MOLE57251431-25GEA 0 EUR
MOLE57251431-25G
(p-Toluolsulfonyl)methylisocyanid
(p-Toluolsulfonyl)methylisocyanid
Formel: C₉H₉NO₂S
Molecular Weight: 195,24 g/mol
MDL: MFCD00000005
CAS-Nummer: 36635-61-7
EINECS: 253-140-1
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR