4-Pyridinboronsäure

Lieferant: Molekula

Synonyme: 4-Pyridinylboronsäure, Pyridin-4-boronsäure

57313082-1G
MOLE57313082-1GEA 0 EUR
MOLE57313082-1G
4-Pyridinboronsäure
4-Pyridineboronic acid
Formel: C₅H₆BNO₂
Molecular Weight: 122,92 g/mol (anhy)
MDL: MFCD01074545
CAS-Nummer: 1692-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR